Proadifen hydrochloride 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Proadifen HCl

45788-0010 45788-0050
TS45788-0010EA 254 USD
TS45788-0010 TS45788-0050
Proadifen hydrochloride 95%
Proadifen hydrochloride
Formula: C₂₃H₃₁NO₂·HCl
MW: 389.964 g/mol
Melting Pt: 122 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00055151
CAS Number: 62-68-0

Order Now

Specification Test Results

Appearance White to light yellow Powder and/or chunks
Infrared spectrum Conforms
Melting point 121 to 123 °C
TLC ≥94 %
Chloride (Cl) 8.6 to 9.6 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR