L-β-Homoproline hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

42617-0010 42617-2500
TS42617-0010EA 500 USD
TS42617-0010 TS42617-2500
L-β-Homoproline hydrochloride 98%
L-beta-Homoproline hydrochloride
Formula: C₆H₁₂ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD07363484
CAS Number: 53912-85-9

Order Now

Specification Test Results

Appearance White to tan Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 to ≤102.5 %
NMR (1H-NMR) Consistent with structure
Specific optical rotation +31° to +35° (20°C, 589 nm) (c=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR