L-Lysine acetate salt 97%

Supplier: Thermo Scientific Chemicals

Synonyms: L-LYSINE, MONOACETATE, (2S)-2,6-diaminohexanoic acid, L-Lysine acetate, L-Lysine Acetate Salt

45163-0250 45163-1000 45163-5000
TS45163-0250EA 79 USD
TS45163-0250 TS45163-1000 TS45163-5000
L-Lysine acetate salt 97%
L-Lysine acetate salt
Formula: C₈H₁₈N₂O₄
MW: 206.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039069
CAS Number: 57282-49-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Assay ≥96.0 %
Specific optical rotation +8° to +10° (20°C, 589 nm) (c=10, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR