N-Boc-4-oxo-L-proline methyl ester 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-1-tert-Butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate, Boc-4-oxo-Pro-OMe

44927-0010 44927-0050
TS44927-0010EA 65 USD
TS44927-0010 TS44927-0050
N-Boc-4-oxo-L-proline methyl ester 97%
N-Boc-4-oxo-L-proline methyl ester
Formula: C₁₁H₁₇NO₅
MW: 243.259 g/mol
Melting Pt: 42…45 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861778
CAS Number: 102195-80-2

Order Now

Specification Test Results

Appearance White to tan Solid
Infrared spectrum Conforms
Melting point 42 to 46 °C
GC ≥96.0 %
Specific optical rotation +10° to +17° (20 °C, 589 nm) (c=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR