Cefadroxil 95-105 %

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cefadroxil monohydrate, (6R,7R)-7-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

45947-0010 45947-0050
TS45947-0010EA 85 USD
TS45947-0010 TS45947-0050
Cefadroxil 95-105 %
Cefadroxil
Formula: C₁₆H₁₇N₃O₅S
MW: 363.388 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00865091
CAS Number: 50370-12-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water 4.2 to 6.0 %
HPLC ≥95 %
pH 4.0 to 6.0
Specific optical rotation 165° to 178° (20°C, 589 nm) (1 % in water DS)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR