N-Acetyl-L-proline 99%

Supplier: Thermo Scientific Chemicals

37664-0010 37664-0050
TS37664-0010EA 49 USD
TS37664-0010 TS37664-0050
N-Acetyl-L-proline 99%
N-Acetyl-L-proline
Formula: C₇H₁₁NO₃
Melting Pt: 115…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00020837
CAS Number: 68-95-1
EINECS: 276-259-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Specific optical rotation -114° to -118° (20°C, 589 nm) (C=1, Water)
Titration with TBAH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR