N-Acetyl-L-tyrosine ethyl ester monohydrate 99%

Supplier: Thermo Scientific Chemicals

10275-0010 10275-0100 10275-0500
TS10275-0010EA 31 USD
TS10275-0010 TS10275-0100 TS10275-0500
N-Acetyl-L-tyrosine ethyl ester monohydrate 99%
N-Acetyl-L-tyrosine ethyl ester monohydrate
Formula: C₁₃H₁₉NO₅
MW: 269.297 g/mol
Melting Pt: 80…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00149093
CAS Number: 36546-50-6

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water 3.5 to 8.0 % (K.F.)
HPLC ≥98.5 %
Appearance of solution (5% soln. in EtOH) clear, colorless
Specific optical rotation +22° to +24° (24°C, 589 nm) (c=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR