Methyl cellulose, 15 cP (2% H₂O)

Supplier: Thermo Scientific Chemicals

Synonyms: Cellulose methyl ether

18231-0010 18231-1000 18231-2500
TS18231-0010EA 233 USD
TS18231-0010 TS18231-1000 TS18231-2500
Methyl cellulose, 15 cP (2% H₂O)
Methyl cellulose
Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5

Order Now

Specification Test Results

Appearance White to yellowish-white to greyish-white Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Viscosity 12 to 18 cP (2 % in water at 20°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR