N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride, BAPNA, BANI, Bz-DL-Arg-pNA·HCl

22774-0010 22774-0050
TS22774-0010EA 95 USD
TS22774-0010 TS22774-0050
N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride 98%
N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride
Formula: C₁₉H₂₃ClN₆O₄
MW: 434.881 g/mol
Melting Pt: 268…272 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012846
CAS Number: 911-77-3

Order Now

Specification Test Results

Appearance White to yellow to light brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR