D-Propargylglycine 98%, 98% ee

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-D-Pra-OH, (R)-2-Aminopent-4-ynoic acid, (R)-alpha-Propargylglycine

44122-1000 44122-5000
TS44122-1000EA 191 USD
TS44122-1000 TS44122-5000
D-Propargylglycine 98%, 98% ee
D-Propargylglycine
Formula: C₅H₇NO₂
Melting Pt: >180 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00237407
CAS Number: 23235-03-2

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Assay 97.5 to 102.5 %
Specific optical rotation +31° to +35° (20°C, 589 nm) (c=1 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR