DL-Propargylglycine 98%

Supplier: Thermo Scientific Chemicals
Warning

43732-0010 43732-0050
TS43732-0010EA 155 USD
TS43732-0010 TS43732-0050
DL-Propargylglycine 98%
DL-Propargylglycine
Formula: C₅H₇NO₂
Melting Pt: 235 °C
Storage Temperature: Freezer
MDL Number: MFCD00056728
CAS Number: 64165-64-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR