(S)-(+)-2-Phenylglycine methyl ester hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

44219-0100 44219-0500
TS44219-0100EA 35 USD
TS44219-0100 TS44219-0500
(S)-(+)-2-Phenylglycine methyl ester hydrochloride 97%
(S)-(+)-2-Phenylglycine methyl ester hydrochloride
Formula: C₉H₁₂ClNO₂
Melting Pt: 200 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00077158
CAS Number: 15028-39-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥96.0 %
Specific optical rotation +131° to +137° (20°C, 589 nm) (c=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR