β-Alanine-tert-butylester hydrochloride 98%

Supplier: Thermo Scientific Chemicals

43465-0010 43465-0050
TS43465-0010EA 125 USD
TS43465-0010 TS43465-0050
β-Alanine-tert-butylester hydrochloride 98%
beta-Alanine-tert-butylester hydrochloride
Formula: C₇H₁₆ClNO₂
Melting Pt: 167.8…168.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00058258
CAS Number: 58620-93-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or solid
Infrared spectrum Conforms
Elemental Analysis 7.5 to 7.9 % N
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR