L-Phenylalanine ethyl ester hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

20543-0100 20543-0500
TS20543-0100EA 62.3 USD
TS20543-0100 TS20543-0500
L-Phenylalanine ethyl ester hydrochloride 99%
L-Phenylalanine ethyl ester hydrochloride
Formula: C₁₁H₁₆ClNO₂
Melting Pt: 154…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00012507
CAS Number: 3182-93-2

Order Now

Specification Test Results

Appearance White Crystalline powder or needles
Infrared spectrum Conforms
Melting point 154 to 157 °C
Specific optical rotation +32.2° to +35.2° (20 °C, 589 nm) (c=2, C2H5OH)
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR