Nα-Fmoc-N-δ-trityl-L-glutamine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Fmoc-Gln(Trt)-OH

44064-0050 44064-0250
TS44064-0050EA 25 USD
TS44064-0050 TS44064-0250
Nα-Fmoc-N-δ-trityl-L-glutamine 95%
N-alpha-Fmoc-N-delta-trityl-L-glutamine
Formula: C₃₉H₃₄N₂O₅
MW: 610.71 g/mol
Melting Pt: 131…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077056
CAS Number: 132327-80-1

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -12.5° to -15.5° (20°C, 589 nm) (c=1 in DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR