N-Boc-S-trityl-L-cysteine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Boc-S-trityl-L-cysteine, N-(tert-Butoxycarbonyl)-S-trityl-L-cysteine, BOC-S-trityl-L-cysteine, (R)-2-((tert-Butoxycarbonyl)amino)-3-(tritylthio)propanoic acid, Boc-Cys(Trt)-OH

39906-0010 39906-0050
TS39906-0010EA 36 USD
TS39906-0010 TS39906-0050
N-Boc-S-trityl-L-cysteine 99%
N-Boc-S-trityl-L-cysteine
Formula: C₂₇H₂₉NO₄S
Melting Pt: 143…146 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038251
CAS Number: 21947-98-8

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 142 to 154 °C
HPLC ≥98.5 %
Specific optical rotation +26° to +30° (20 °C, 589 nm) (c=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR