Boc-sarcosine 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-Sar-OH

27567-0050 27567-0250
TS27567-0050EA 31 USD
TS27567-0050 TS27567-0250
Boc-sarcosine 99+%
Boc-sarcosine
Formula: C₈H₁₅NO₄
MW: 189.211 g/mol
Melting Pt: 86…90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037795
CAS Number: 13734-36-6

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 86 to 90 °C
Water ≤0.2 % (K.F.)
HPLC ≥99.0 % (ELSD)
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR