Zonisamide

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Benzisoxazole-3-methanesulphonamide, Benzo[d]isoxazol-3-ylmethanesulphonamide

46504-0010 46504-2500
TS46504-0010EA 251 USD
TS46504-0010 TS46504-2500
Zonisamide
Zonisamide
Formula: C₈H₈N₂O₃S
MW: 212.22 g/mol
Melting Pt: 165 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00865316
CAS Number: 68291-97-4

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 163 to 167 °C
Water ≤1 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR