Yohimbine hydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
14103-0050
14103-0250
TS14103-0050EA
104
USD
TS14103-0050
TS14103-0250
Yohimbine hydrochloride 99%
Yohimbine hydrochloride
Formula:
C₂₁H₂₆N₂O₃ MW: 390.91 g/mol Melting Pt: 285…288 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012674 CAS Number: 65-19-0 UN: 2811 ADR: 6.1 II |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Specific optical rotation | +99° to +105° (20°C, 589 nm) (c=1, H2O) |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...