Xylometazoline hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Danger

46199-0050 46199-0250
TS46199-0050EA 111 USD
TS46199-0050 TS46199-0250
Xylometazoline hydrochloride 98%
Xylometazoline hydrochloride
Formula: C₁₆H₂₄N₂
Storage Temperature: Ambient
MDL Number: MFCD00238707
CAS Number: 1218-35-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 4 h)
Solubility (5 % in water) Clear colorless to light yellow
Titration with HClO4 99 to 101 % (dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR