Xanthosine dihydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 9-β-D-Ribofuranosylxanthine dihydrate, 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purine-2,6-diol dihydrate, 3,9-Dihydro-9-beta-D-ribofuranosyl-1H-purine-2,6-dione dihydrate

20763-0010 20763-0050
TS20763-0010EA 60 USD
TS20763-0010 TS20763-0050
Xanthosine dihydrate 99%
Xanthosine dihydrate
Formula: C₁₀H₁₂N₄O₆·2H₂O
MW: 320.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00149347
CAS Number: 5968-90-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water 9 to 14 %
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR