Verapamil hydrochloride 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-((3,4-Dimethoxyphenethyl)(methyl)amino)-2-(3,4-dimethoxyphenyl)-2-isopropylpentanenitrile hydrochloride, Verapamil HCl, 5-[(3,4-Dimethoxyphenethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile Hydrochloride, (±)-Verapamil hydrochloride
32933-0010
32933-0050
TS32933-0010EA
113
USD
TS32933-0010
TS32933-0050
Verapamil hydrochloride 99+%
Verapamil hydrochloride
Formula:
C₂₇H₃₈N₂O₄ MW: 491.07 g/mol Melting Pt: 140…144 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00055208 CAS Number: 152-11-4 UN: 2811 ADR: 6.1 III |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 140 to 144 °C |
Loss on drying | ≤0.5 % (105 °C, 2 h) |
Residue after ignition | ≤0.1 % |
pH | 4.5 to 6.5 (5 % solution) |
Titration with HClO4 | 99.0 to 100.5 % (on dried substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...