Trimethoprim 98%

Supplier: Thermo Scientific Chemicals
Warning

45512-0050 45512-0250 45512-1000
TS45512-0050EA 114 USD
TS45512-0050 TS45512-0250 TS45512-1000
Trimethoprim 98%
Trimethoprim
Formula: C₁₄H₁₈N₄O₃
Melting Pt: 234…236 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036761
CAS Number: 738-70-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 199 to 203 °C
Loss on drying ≤0.5 % (105 °C, 4 h)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR