Triamcinolone
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
9alpha-Fluoro-11beta,16alpha,17,21-tetrahydroxy-1,4-pregnadiene-3,20-dione, Fluoxyprednisolone
46224-0010
46224-0050
TS46224-0010EA
106
USD
TS46224-0010
TS46224-0050
Triamcinolone
Triamcinolone
Formula:
C₂₁H₂₇FO₆ MW: 394.44 g/mol Melting Pt: 262…263 °C Storage Temperature: Refrigerator |
CAS Number:
124-94-7 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...