Tinidazole

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-[2-(Ethylsulphonyl)ethyl]-2-methyl-5-nitroimidazole, 1-(2-(Ethylsulphonyl)ethyl)-2-methyl-5-nitro-1H-imidazole

46351-0050 46351-0250
TS46351-0050EA 36 USD
TS46351-0050 TS46351-0250
Tinidazole
Tinidazole
Formula: C₈H₁₃N₃O₄S
MW: 247.27 g/mol
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00057217
CAS Number: 19387-91-8

Order Now

Specification Test Results

Appearance Almost white to light yellow Powder
Infrared spectrum Conforms
Melting point 125 to 128 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR