Thiamphenicol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2-Dichloro-N-[(1R,2R)-1,3-dihydroxy-1-[4-(methylsulphonyl)phenyl]-2-propyl]acetamide, (Methylsulphonyl)chloramphenicol

45545-0010 45545-0050 45545-0250
TS45545-0010EA 27 USD
TS45545-0010 TS45545-0050 TS45545-0250
Thiamphenicol 98%
Thiamphenicol
Formula: C₁₂H₁₅Cl₂NO₅S
MW: 356.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00467983
CAS Number: 15318-45-3

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 160 to 170 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR