Sulbactam

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sulbactam acid, (2S,5R)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid 4,4-Dioxide

45535-0050 45535-0250
TS45535-0050EA 97 USD
TS45535-0050 TS45535-0250
Sulbactam
Sulbactam
Formula: C₈H₁₁NO₅S
MW: 233.24 g/mol
Melting Pt: 156 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00867005
CAS Number: 68373-14-8

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 146 to 151 °C
Water ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR