Stachyose hydrate, Technical Grade

Supplier: Thermo Scientific Chemicals

Synonyms: Manneotetrose, beta-D-Fructofuranosyl O-alpha-D-Galactopyranosyl-(1-6)-O-alpha-D-galactopyranosyl-(1-6)-alpha-D-glucopyranoside, Stachyose hydrate(1:x), Lupeose

45314-0010 45314-0050 45314-2500
TS45314-0010EA 215 USD
TS45314-0010 TS45314-0050 TS45314-2500
Stachyose hydrate, Technical Grade
Stachyose hydrate
Formula: C₂₄H₄₂O₂₁·xH₂O
MW: 666.58 g/mol (anhy)
Melting Pt: 95…105 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149457
CAS Number: 54261-98-2

Order Now

Specification Test Results

Appearance White to light yellow to light beige Powder
Infrared spectrum Conforms
Water ≤12 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR