Stachyose hydrate 97+%

Supplier: Thermo Scientific Chemicals

Synonyms: Manneotetrose, beta-D-Fructofuranosyl O-alpha-D-Galactopyranosyl-(1-6)-O-alpha-D-galactopyranosyl-(1-6)-alpha-D-glucopyranoside, Stachyose hydrate(1:x), Lupeose

22608-0010 22608-0050 22608-5000
TS22608-0010EA 251 USD
TS22608-0010 TS22608-0050 TS22608-5000
Stachyose hydrate 97+%
Stachyose hydrate
Formula: C₂₄H₄₂O₂₁·xH₂O
MW: 666.58 g/mol (anhy)
Melting Pt: 95…105 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149457
CAS Number: 54261-98-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤12 % (K.F.)
Solubility (5 % in water) Clear to slightly hazy colorless
HPLC ≥97.0 % (ELSD)
Specific optical rotation +129° to +140° (20°C, 589 nm) (c=4, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR