Prasugrel 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-[2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl Acetate, 5-(2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate

46148-0010 46148-2500
TS46148-0010EA 157 USD
TS46148-0010 TS46148-2500
Prasugrel 98%
Prasugrel
Formula: C₂₀H₂₀FNO₃S
MW: 373.44 g/mol
Melting Pt: 110…121 °C
Storage Temperature: Freezer
MDL Number: MFCD09954140
CAS Number: 150322-43-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥97.5 %
Residue after ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR