Pentasodium diethylenetriaminepentaacetate 40% in aqueous solution tech.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diethylenetriaminepentaacetic Acid Pentasodium Salt

40729-0010 40729-0030 40729-2500
TS40729-0010EA 157 USD
TS40729-0010 TS40729-0030 TS40729-2500
Pentasodium diethylenetriaminepentaacetate 40% in aqueous solution tech.
Pentasodium diethylenetriaminepentaacetate
Formula: C₁₄H₂₃N₃O₁₀
MW: 503.26 g/mol
Boiling Pt: 106 °C (760 mmHg)
Melting Pt: –40 °C
Density: 1.29 g/cm³
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00051016
CAS Number: 140-01-2
UN: 3267

Order Now

Specification Test Results

Appearance Clear solution
Color scale ≤150 APHA
pH 11.0 to 12.0 (1 % in water)
Titration 39 to 41 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR