Penta-O-acetyl-β-D-glucopyranose 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
β-D-Glucose Pentaacetate, Pentaacetyl-β-D-glucose, B-D-Glucose Pentaacetate, (2S,3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate
30296-1000
30296-5000
TS30296-1000EA
67
USD
TS30296-1000
TS30296-5000
Penta-O-acetyl-β-D-glucopyranose 98%
Penta-O-acetyl-beta-D-glucopyranose
Formula:
C₁₆H₂₂O₁₁ Melting Pt: 129…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006597 CAS Number: 604-69-3 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % (K.F.) |
Sulfated ash | ≤0.5 % |
GC | ≥97.5 % |
Specific optical rotation | +4° to +6° (20°C, 589 nm) (c=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...