Parthenolide

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,5alpha-Epoxy-6beta-hydroxygermacra-1(10),11(13)-dien-12-oic Acid gamma-Lactone, (3aS,9aR,10aR,10bS,E)-6,9a-Dimethyl-3-methylene-3a,4,5,8,9,9a,10a,10b-octahydrooxireno[2',3':9,10]cyclodeca[1,2-b]furan-2(3H)-one

46215-1000 46215-5000
TS46215-1000EA 157 USD
TS46215-1000 TS46215-5000
Parthenolide
Parthenolide
Formula: C₁₅H₂₀O₃
MW: 248.32 g/mol
Melting Pt: 115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134592
CAS Number: 20554-84-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 113 to 117 °C
Solubility (5% in CHCl3) Clear solution
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR