Ofloxacin 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid, 9-Fluoro-3,7-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid

45567-0010 45567-0050 45567-0250
TS45567-0010EA 53 USD
TS45567-0010 TS45567-0050 TS45567-0250
Ofloxacin 98%
Ofloxacin
Formula: C₁₈H₂₀FN₃O₄
MW: 361.37 g/mol
Melting Pt: 280…275 °C
Storage Temperature: Freezer
MDL Number: MFCD00226105
CAS Number: 82419-36-1

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Loss on drying ≤0.2 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR