Naftifine hydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Naftifine HCl, (E)-N-methyl-N-(naphthalen-1-ylmethyl)-3-phenylprop-2-en-1-amine hydrochloride, AW 105843, SN 105843, N-trans-Cinnamyl-N-methyl-(1-naphthylmethyl)amine hydrochloride

45866-0010 45866-0050 458660250
TS45866-0010EA 89 USD
TS45866-0010 TS45866-0050 TS458660250
Naftifine hydrochloride 99%
Naftifine hydrochloride
Formula: C₂₁H₂₁N
MW: 323.86 g/mol
Melting Pt: 175…179 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00059047
CAS Number: 65473-14-5

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 174 to 180 °C
Titration with HClO4 ≥98.5 %
Loss on fusion ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR