N-Succinimidyl Acetate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Acetoxysuccinimide, Ac-Osu, Acetic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl acetate

46475-0050 464750250 464751000
TS46475-0050EA 63 USD
TS46475-0050 TS464750250 TS464751000
N-Succinimidyl Acetate
N-Succinimidyl Acetate
Formula: C₆H₇NO₄
MW: 157.13 g/mol
Melting Pt: 133…134 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037916
CAS Number: 14464-29-0

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 132 to 136 °C
Water ≤2 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR