Nω-Nitro-L-arginine methyl ester hydrochloride 98%
Supplier: Thermo Scientific Chemicals
32875-0050
32875-0250
TS32875-0050EA
153
USD
TS32875-0050
TS32875-0250
Nω-Nitro-L-arginine methyl ester hydrochloride 98%
Nomega-Nitro-L-arginine methyl ester hydrochloride
Formula:
C₇H₁₅N₅O₄ Melting Pt: 159…161 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00039052 CAS Number: 51298-62-5 |
Specification Test Results
Appearance | White to off-white Crystalline solid |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Specific optical rotation | +14.0° to +16.0° (24°C, 589 nm) (c=3, CH3OH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...