N-Methyl-D-aspartic acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Methyl-D-asparaginic Acid, NMDA, Me-D-Asp-OH, (R)-2-(Methylamino)succinic acid
32919-0500
32919-2500
TS32919-0500EA
82
USD
TS32919-0500
TS32919-2500
N-Methyl-D-aspartic acid 98%
N-Methyl-D-aspartic acid
Formula:
C₅H₉NO₄ Melting Pt: 187…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004226 CAS Number: 6384-92-5 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Specific optical rotation | -26° to -30° (20°C, 589 nm) (c=1, 6 N HCl) |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...