N-Isopropylacrylamide 99% stabilized, pure

Supplier: Thermo Scientific Chemicals
Warning

41278-5000 41278-0025 41278-0250 41278-1000
TS41278-5000EA 799 USD
TS41278-5000 TS41278-0025 TS41278-0250 TS41278-1000
N-Isopropylacrylamide 99% stabilized, pure
N-Isopropylacrylamide
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 89…92 °C (2 mmHg)
Melting Pt: 62…66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041913
CAS Number: 2210-25-5

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 62 to 66 °C
GC ≥98.5 %
Stabilizer ca. 500 ppm MEHQ

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR