Nifedipine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Adalat, 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic Acid Dimethyl Ester
32927-0010
32927-0050
32927-0250
TS32927-0010EA
68
USD
TS32927-0010
TS32927-0050
TS32927-0250
Nifedipine 98+%
Nifedipine
Formula:
C₁₇H₁₈N₂O₆ MW: 346.34 g/mol Melting Pt: 171…175 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00057326 CAS Number: 21829-25-4 |
Specification Test Results
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
Melting point | 171 to 175 °C |
Loss on drying | ≤0.5 % |
HPLC | ≥98.0 % |
Residue after ignition | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...