Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Bz-Arg-OEt.HCl, Nα-Benzoyl-L-arginine Ethyl Ester HCl

10570-0100 10570-0025 10570-0500
TS10570-0100EA 123 USD
TS10570-0100 TS10570-0025 TS10570-0500
Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride 99+%
Nalpha-Benzoyl-L-arginine ethyl ester hydrochloride
Formula: C₁₅H₂₂N₄O₃
MW: 342.82 g/mol
Melting Pt: 129…136 °C
Storage Temperature: Freezer
MDL Number: MFCD00012579
CAS Number: 2645-08-1

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless
TLC ≥99.0 %
Specific optical rotation -15.0° to -19.0° (24°C, 589 nm) (c=2, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR