Nɑ-Acetyl-L-lysine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Nalpha-Acetyl-L-lysine, Ac-Lys-OH

22763-0010 22763-2500
TS22763-0010EA 107 USD
TS22763-0010 TS22763-2500
Nɑ-Acetyl-L-lysine 99+%
Nalpha-Acetyl-L-lysine
Formula: C₈H₁₆N₂O₃
Melting Pt: 256…258 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008233
CAS Number: 1946-82-3

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Specific optical rotation +5.0° to +6.0° (20°C, 589 nm) (c=1, H2O)
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR