FMOC-O-tert-butyl-L-tyrosine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-α-Fmoc-o-t-butyl-L-Tyrosine, N-Fmoc-O-tert-butyl-L-tyrosine, Nalpha-Fmoc-O-tert-butyl-L-tyrosine, O-tert-Butyl-Nalpha-Fmoc-L-tyrosine, Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-tyrosine, Fmoc-Tyr(tBu)-OH

43909-0100 43909-0250
TS43909-0100EA 42 USD
TS43909-0100 TS43909-0250
FMOC-O-tert-butyl-L-tyrosine 98%
FMOC-O-tert-butyl-L-tyrosine
Formula: C₂₈H₂₉NO₅
Melting Pt: 150…156 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037129
CAS Number: 71989-38-3

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -27° to -31° (20°C, 589 nm) (c=1 in DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR