N-(tert-Butoxycarbonyl)-L-tert-leucine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Boc-L-tert-leucine, Boc-Tle-OH, (S)-2-(Boc-amino)-3,3-dimethylbutyric Acid, (S)-2-(tert-Butoxycarbonylamino)-3,3-dimethylbutyric Acid

36801-0010 36801-0050 36801-0250
TS36801-0010EA 40 USD
TS36801-0010 TS36801-0050 TS36801-0250
N-(tert-Butoxycarbonyl)-L-tert-leucine 98%
N-(tert-Butoxycarbonyl)-L-tert-leucine
Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol
Melting Pt: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00065574
CAS Number: 62965-35-9

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Specific optical rotation -3.0° to -5.0° (20°C, 589 nm) (c=1, acetic acid)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR