N-(tert-Butoxycarbonyl)-L-2-phenylglycine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-Phg-OH, BOC-L-alpha-phenylglycine, Boc-D-Phg-OH, N-Boc-L-2-phenylglycine

39816-0010 39816-0050
TS39816-0010EA 43 USD
TS39816-0010 TS39816-0050
N-(tert-Butoxycarbonyl)-L-2-phenylglycine 99%
N-(tert-Butoxycarbonyl)-L-2-phenylglycine
Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00065588
CAS Number: 2900-27-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 88 to 91 °C
Specific optical rotation +142° to +146° (20 °C, 589 nm) (c=1%, ethanol)
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR