N-(tert-Butoxycarbonyl)-D-phenylalanine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Boc-D-phenylalanine, BOC-D-Phenylalanine, Boc-D-Phe-OH

27574-0050 27574-0250
TS27574-0050EA 64 USD
TS27574-0050 TS27574-0250
N-(tert-Butoxycarbonyl)-D-phenylalanine 99+%
N-(tert-Butoxycarbonyl)-D-phenylalanine
Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
Melting Pt: 78…87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063149
CAS Number: 18942-49-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 85 to 88 °C
Water ≤0.5 % (K.F.)
HPLC ≥99.0 %
Specific optical rotation -24° to -26° (20 °C, 589 nm) (c=1, ethanol)
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR