N-(tert-Butoxycarbonyl)-D-2-phenylglycine 99%

Supplier: Thermo Scientific Chemicals
Warning

43764-0010 43764-0050
TS43764-0010EA 44 USD
TS43764-0010 TS43764-0050
N-(tert-Butoxycarbonyl)-D-2-phenylglycine 99%
N-(tert-Butoxycarbonyl)-D-2-phenylglycine
Formula: C₁₃H₁₇NO₄
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00062043
CAS Number: 33125-05-2

Order Now

Specification Test Results

Appearance White Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation -142° to -146° (20°C, 589 nm) (c=1, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR