N-(tert-Butoxycarbonyl)-4-aminobutyric acid 98%

Supplier: Thermo Scientific Chemicals

37657-0010 37657-0050 37657-0250
TS37657-0010EA 48 USD
TS37657-0010 TS37657-0050 TS37657-0250
N-(tert-Butoxycarbonyl)-4-aminobutyric acid 98%
N-(tert-Butoxycarbonyl)-4-aminobutyric acid
Formula: C₉H₁₇NO₄
Boiling Pt: 78…82 °C (.2 mmHg)
Melting Pt: 55 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00037313
CAS Number: 57294-38-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 54 to 60 °C
TLC ≥97.5 %
Titration with NaOH 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR