N-(3-Trifluoromethylphenyl)anthranilic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Flufenamic acid, 2-[(3-Trifluoromethylphenyl)amino]benzoic acid, 2-(3-Trifluoromethylanilino)benzoic acid, 2-((3-(Trifluoromethyl)phenyl)amino)benzoic acid

16592-0100 16592-0500
TS16592-0100EA 28 USD
TS16592-0100 TS16592-0500
N-(3-Trifluoromethylphenyl)anthranilic acid 97%
N-(3-Trifluoromethylphenyl)anthranilic acid
Formula: C₁₄H₁₀F₃NO₂
MW: 281.23 g/mol
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00002422
CAS Number: 530-78-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow to grey-green Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 132 to 137 °C
Titration with NaOH 96.0 to 104.0 %
Residue after ignition ≤0.5 %
Specific optical rotation -91.0° to -93.5° (20 °C, 589 nm)
Acidity Passes test
Color of solution Passes test
Chloride (Cl) ≤0.018 %
Sulfate (SO4) ≤0.025 %
Arsenic (As) ≤1 ppm
Ca and Mg ≤0.005 % (as Ca)
Hydroxymethylfurfural Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR