L-Tyrosine methyl ester 98%

Supplier: Thermo Scientific Chemicals
Warning

14065-0050 14065-0250
TS14065-0050EA 44 USD
TS14065-0050 TS14065-0250
L-Tyrosine methyl ester 98%
L-Tyrosine methyl ester
Formula: C₁₀H₁₃NO₃
Melting Pt: 133…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002392
CAS Number: 1080-06-4

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %
Appearance of solution (5% soln. in methanol) Clear to slightly hazy
Specific optical rotation +23° to +29° (20°C, 589 nm) (c=2.4, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR